Anti-CXCL8 Rat Monoclonal Antibody [clone: 344609]

Supplier: R & D Systems
MAB2277
220019-735EA 561.11 USD
220019-735
Anti-CXCL8 Rat Monoclonal Antibody [clone: 344609]
Antibodies
Type: Primary
Antigen: CXCL8
Clonality: Monoclonal
Clone: 344609
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR