Anti-IL17RD Rat Monoclonal Antibody [clone: 400210]

Supplier: R & D Systems
MAB2276
220019-734EA 577.02 USD
220019-734
Anti-IL17RD Rat Monoclonal Antibody [clone: 400210]
Antibodies
Type: Primary
Antigen: IL17RD
Clonality: Monoclonal
Clone: 400210
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR