Anti-IL17RD Mouse Monoclonal Antibody [clone: 309511]

Supplier: R & D Systems
MAB22751
102885-448EA 594.37 USD
102885-448
Anti-IL17RD Mouse Monoclonal Antibody [clone: 309511]
Antibodies
Type: Primary
Antigen: IL17RD
Clonality: Monoclonal
Clone: 309511
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR