Anti-IL17RD Mouse Monoclonal Antibody [clone: 309539]

Supplier: R & D Systems
MAB2275
220019-733EA 594.37 USD
220019-733
Anti-IL17RD Mouse Monoclonal Antibody [clone: 309539]
Antibodies
Type: Primary
Antigen: IL17RD
Clonality: Monoclonal
Clone: 309539
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR