Anti-IL17D Rat Monoclonal Antibody [clone: 312724]

Supplier: R & D Systems
MAB2274
220019-732EA 594.37 USD
220019-732
Anti-IL17D Rat Monoclonal Antibody [clone: 312724]
Antibodies
Type: Primary
Antigen: IL17D
Clonality: Monoclonal
Clone: 312724
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR