Anti-IL6R Mouse Monoclonal Antibody [clone: 556704]

Supplier: R & D Systems
MAB2271
220019-731EA 594.37 USD
220019-731
Anti-IL6R Mouse Monoclonal Antibody [clone: 556704]
Antibodies
Type: Primary
Antigen: IL6R
Clonality: Monoclonal
Clone: 556704
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR