Anti-IL17RC Mouse Monoclonal Antibody [clone: 309822]

Supplier: R & D Systems
MAB22691
220019-729EA 594.37 USD
220019-729
Anti-IL17RC Mouse Monoclonal Antibody [clone: 309822]
Antibodies
Type: Primary
Antigen: IL17RC
Clonality: Monoclonal
Clone: 309822
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR