Anti-KITLG Mouse Monoclonal Antibody [clone: 371907]

Supplier: R & D Systems
MAB2268
220019-728EA 577.02 USD
220019-728
Anti-KITLG Mouse Monoclonal Antibody [clone: 371907]
Antibodies
Type: Primary
Antigen: KITLG
Clonality: Monoclonal
Clone: 371907
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR