Anti-FAS Mouse Monoclonal Antibody [clone: 431006]

Supplier: R & D Systems
MAB22671
220019-727EA 561.11 USD
220019-727
Anti-FAS Mouse Monoclonal Antibody [clone: 431006]
Antibodies
Type: Primary
Antigen Symbol: FAS
Antigen Name:
Antigen Synonyms: Apo-1, APT1, CD95, TNFRSF6
Clonality: Monoclonal
Clone: 431006
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR