Anti-SEMA6B Rat Monoclonal Antibody [clone: 256537]

Supplier: R & D Systems
MAB2264
220019-724EA 763.57 USD
220019-724
Anti-SEMA6B Rat Monoclonal Antibody [clone: 256537]
Antibodies
Type: Primary
Antigen: SEMA6B
Clonality: Monoclonal
Clone: 256537
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR