Anti-BMP2 Mouse Monoclonal Antibody [clone: 253717]

Supplier: R & D Systems
MAB2260
220019-722EA 763.57 USD
220019-722
Anti-BMP2 Mouse Monoclonal Antibody [clone: 253717]
Antibodies
Type: Primary
Antigen: BMP2
Clonality: Monoclonal
Clone: 253717
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR