Anti-CTSL Mouse Monoclonal Antibody [clone: 193702]

Supplier: R & D Systems
MAB22591
220019-720EA 594.37 USD
220019-720
Anti-CTSL Mouse Monoclonal Antibody [clone: 193702]
Antibodies
Type: Primary
Antigen: CTSL
Clonality: Monoclonal
Clone: 193702
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR