Anti-CTSL Mouse Monoclonal Antibody [clone: 193720]

Supplier: R & D Systems
MAB2259
220019-719EA 772.25 USD
220019-719
Anti-CTSL Mouse Monoclonal Antibody [clone: 193720]
Antibodies
Type: Primary
Antigen: CTSL
Clonality: Monoclonal
Clone: 193720
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR