Anti-CDKN1B Rat Monoclonal Antibody [clone: 341116]

Supplier: R & D Systems
MAB22561
220019-717EA 594.37 USD
220019-717
Anti-CDKN1B Rat Monoclonal Antibody [clone: 341116]
Antibodies
Type: Primary
Antigen: CDKN1B
Clonality: Monoclonal
Clone: 341116
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR