Anti-CDKN1B Mouse Monoclonal Antibody [clone: 225501]

Supplier: R & D Systems
MAB2256
220019-716EA 594.37 USD
220019-716
Anti-CDKN1B Mouse Monoclonal Antibody [clone: 225501]
Antibodies
Type: Primary
Antigen: CDKN1B
Clonality: Monoclonal
Clone: 225501
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR