Anti-LDLR Rat Monoclonal Antibody [clone: 263123]

Supplier: R & D Systems
MAB2255
220019-715EA 594.37 USD
220019-715
Anti-LDLR Rat Monoclonal Antibody [clone: 263123]
Antibodies
Type: Primary
Antigen: LDLR
Clonality: Monoclonal
Clone: 263123
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR