Anti-DNER Rat Monoclonal Antibody [clone: 298306]

Supplier: R & D Systems
MAB2254
220019-714EA 561.11 USD
220019-714
Anti-DNER Rat Monoclonal Antibody [clone: 298306]
Antibodies
Type: Primary
Antigen: DNER
Clonality: Monoclonal
Clone: 298306
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR