Anti-NCR2 Mouse Monoclonal Antibody [clone: 253422]

Supplier: R & D Systems
MAB2249
220019-711EA 577.02 USD
220019-711
Anti-NCR2 Mouse Monoclonal Antibody [clone: 253422]
Antibodies
Type: Primary
Antigen: NCR2
Clonality: Monoclonal
Clone: 253422
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR