Anti-PROCR Mouse Monoclonal Antibody [clone: 304519]

Supplier: R & D Systems
MAB22451
220019-709EA 594.37 USD
220019-709
Anti-PROCR Mouse Monoclonal Antibody [clone: 304519]
Antibodies
Type: Primary
Antigen: PROCR
Clonality: Monoclonal
Clone: 304519
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR