Anti-CEACAM1 Mouse Monoclonal Antibody [clone: 283340]

Supplier: R & D Systems
MAB2244
220019-706EA 639.2 USD
220019-706
Anti-CEACAM1 Mouse Monoclonal Antibody [clone: 283340]
Antibodies
Type: Primary
Antigen: CEACAM1
Clonality: Monoclonal
Clone: 283340
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR