Anti-IL2RB Mouse Monoclonal Antibody [clone: 27311]

Supplier: R & D Systems
MAB2241
220019-705EA 763.57 USD
220019-705
Anti-IL2RB Mouse Monoclonal Antibody [clone: 27311]
Antibodies
Type: Primary
Antigen: IL2RB
Clonality: Monoclonal
Clone: 27311
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR