Anti-CDH12 Rat Monoclonal Antibody [clone: 343621]

Supplier: R & D Systems
MAB2240
220019-704EA 577.02 USD
220019-704
Anti-CDH12 Rat Monoclonal Antibody [clone: 343621]
Antibodies
Type: Primary
Antigen: CDH12
Clonality: Monoclonal
Clone: 343621
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR