Anti-PCOLCE Rat Monoclonal Antibody [clone: 261845]

Supplier: R & D Systems
MAB2239
220019-702EA 772.25 USD
220019-702
Anti-PCOLCE Rat Monoclonal Antibody [clone: 261845]
Antibodies
Type: Primary
Antigen: PCOLCE
Clonality: Monoclonal
Clone: 261845
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR