Anti-KIR2DL4 Mouse Monoclonal Antibody [clone: 181703]

Supplier: R & D Systems
MAB2238
220019-701EA 779.48 USD
220019-701
Anti-KIR2DL4 Mouse Monoclonal Antibody [clone: 181703]
Antibodies
Agonist Activity

Type: Primary
Antigen: KIR2DL4
Clonality: Monoclonal
Clone: 181703
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR