Anti-SERPINI2 Mouse Monoclonal Antibody [clone: 203402]

Supplier: R & D Systems
MAB2235
220019-699EA 763.57 USD
220019-699
Anti-SERPINI2 Mouse Monoclonal Antibody [clone: 203402]
Antibodies
Type: Primary
Antigen: SERPINI2
Clonality: Monoclonal
Clone: 203402
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR