Anti-SERPINA5 Rat Monoclonal Antibody [clone: 258712]

Supplier: R & D Systems
MAB2234
220019-698EA 763.57 USD
220019-698
Anti-SERPINA5 Rat Monoclonal Antibody [clone: 258712]
Antibodies
Type: Primary
Antigen: SERPINA5
Clonality: Monoclonal
Clone: 258712
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR