Anti-OLIG1/2/3 Mouse Monoclonal Antibody [clone: 257224]

Supplier: R & D Systems
MAB2230
220019-697EA 642.09 USD
220019-697
Anti-OLIG1/2/3 Mouse Monoclonal Antibody [clone: 257224]
Antibodies
Type: Primary
Antigen: OLIG1/2/3
Clonality: Monoclonal
Clone: 257224
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR