Anti-NCR1 Rat Monoclonal Antibody [clone: 29A1.4]

Supplier: R & D Systems
MAB22252
102885-446EA 577.02 USD
102885-446
Anti-NCR1 Rat Monoclonal Antibody [clone: 29A1.4]
Antibodies
Type: Primary
Antigen Symbol: NCR1
Antigen Name: natural cytotoxicity triggering receptor 1
Antigen Synonyms: Ly94, NKp46
Clonality: Monoclonal
Clone: 29A1.4
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR