Anti-NCR1 Rat Monoclonal Antibody [clone: 259018]

Supplier: R & D Systems
MAB2225
220019-695EA 594.37 USD
220019-695
Anti-NCR1 Rat Monoclonal Antibody [clone: 259018]
Antibodies
Type: Primary
Antigen: NCR1
Clonality: Monoclonal
Clone: 259018
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR