Anti-TREM3 Rat Monoclonal Antibody [clone: 295502]

Supplier: R & D Systems
MAB2224
220019-694EA 577.02 USD
220019-694
Anti-TREM3 Rat Monoclonal Antibody [clone: 295502]
Antibodies
Type: Primary
Antigen: TREM3
Clonality: Monoclonal
Clone: 295502
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR