Anti-ACVR1B Mouse Monoclonal Antibody [clone: 54431]

Supplier: R & D Systems
MAB2221
220019-693EA 633.42 USD
220019-693
Anti-ACVR1B Mouse Monoclonal Antibody [clone: 54431]
Antibodies
Type: Primary
Antigen: ACVR1B
Clonality: Monoclonal
Clone: 54431
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR