Anti-ACVR1B Mouse Monoclonal Antibody [clone: 126822]

Supplier: R & D Systems
MAB222
220019-692EA 772.25 USD
220019-692
Anti-ACVR1B Mouse Monoclonal Antibody [clone: 126822]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: ACVR1B
Clonality: Monoclonal
Clone: 126822
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR