Anti-SERPINB5 Rat Monoclonal Antibody [clone: 305339]

Supplier: R & D Systems
MAB2218
220019-690EA 577.02 USD
220019-690
Anti-SERPINB5 Rat Monoclonal Antibody [clone: 305339]
Antibodies
Type: Primary
Antigen: SERPINB5
Clonality: Monoclonal
Clone: 305339
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR