Anti-NEFL Mouse Monoclonal Antibody [clone: 268014]

Supplier: R & D Systems
MAB2216
220019-689EA 600.15 USD
220019-689
Anti-NEFL Mouse Monoclonal Antibody [clone: 268014]
Antibodies
Type: Primary
Antigen Symbol: NEFL
Antigen Name: neurofilament, light polypeptide
Antigen Synonyms: CMT1F, CMT2E, NF-L, NF68, NFL, PPP1R110
Clonality: Monoclonal
Clone: 268014
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR