Anti-NRP2 Mouse Monoclonal Antibody [clone: 257103]

Supplier: R & D Systems
MAB22151
220019-688EA 594.37 USD
220019-688
Anti-NRP2 Mouse Monoclonal Antibody [clone: 257103]
Antibodies
Type: Primary
Antigen: NRP2
Clonality: Monoclonal
Clone: 257103
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR