Anti-FOSB Mouse Monoclonal Antibody [clone: 269928]

Supplier: R & D Systems
MAB2214
220019-686EA 577.02 USD
220019-686
Anti-FOSB Mouse Monoclonal Antibody [clone: 269928]
Antibodies
Type: Primary
Antigen Symbol: FOSB
Antigen Name: FBJ murine osteosarcoma viral oncogene homolog B
Antigen Synonyms: AP-1, G0S3, GOS3, GOSB
Clonality: Monoclonal
Clone: 269928
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR