Anti-MBL2 Rat Monoclonal Antibody [clone: 272801]

Supplier: R & D Systems
MAB2208
220019-681EA 763.57 USD
220019-681
Anti-MBL2 Rat Monoclonal Antibody [clone: 272801]
Antibodies
Type: Primary
Antigen: MBL2
Clonality: Monoclonal
Clone: 272801
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR