Anti-KNG1 Rat Monoclonal Antibody [clone: 288424]

Supplier: R & D Systems
MAB22061
220019-680EA 594.37 USD
220019-680
Anti-KNG1 Rat Monoclonal Antibody [clone: 288424]
Antibodies
Type: Primary
Antigen: KNG1
Clonality: Monoclonal
Clone: 288424
Conjugation: Unconjugated
Epitope: Light Chain
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR