Anti-KNG1 Rat Monoclonal Antibody [clone: 288422]

Supplier: R & D Systems
MAB2206
220019-679EA 577.02 USD
220019-679
Anti-KNG1 Rat Monoclonal Antibody [clone: 288422]
Antibodies
Type: Primary
Antigen: KNG1
Clonality: Monoclonal
Clone: 288422
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR