Anti-CNTN4 Mouse Monoclonal Antibody [clone: 296015]

Supplier: R & D Systems
MAB2205
220019-678EA 577.02 USD
220019-678
Anti-CNTN4 Mouse Monoclonal Antibody [clone: 296015]
Antibodies
Type: Primary
Antigen: CNTN4
Clonality: Monoclonal
Clone: 296015
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR