Anti-ANGPTL1 Mouse Monoclonal Antibody [clone: 25812]

Supplier: R & D Systems
MAB2204
220019-677EA 763.57 USD
220019-677
Anti-ANGPTL1 Mouse Monoclonal Antibody [clone: 25812]
Antibodies
Type: Primary
Antigen: ANGPTL1
Clonality: Monoclonal
Clone: 25812
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR