Anti-AZU1 Mouse Monoclonal Antibody [clone: 246322]

Supplier: R & D Systems
MAB2200
220019-675EA 763.57 USD
220019-675
Anti-AZU1 Mouse Monoclonal Antibody [clone: 246322]
Antibodies
Type: Primary
Antigen: AZU1
Clonality: Monoclonal
Clone: 246322
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR