Anti-LGMN Mouse Monoclonal Antibody [clone: 312109]

Supplier: R & D Systems
MAB21992
220019-673EA 763.57 USD
220019-673
Anti-LGMN Mouse Monoclonal Antibody [clone: 312109]
Antibodies
Type: Primary
Antigen: LGMN
Clonality: Monoclonal
Clone: 312109
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR