Anti-LGMN Mouse Monoclonal Antibody [clone: 312114]

Supplier: R & D Systems
MAB2199
220019-672EA 577.02 USD
220019-672
Anti-LGMN Mouse Monoclonal Antibody [clone: 312114]
Antibodies
Type: Primary
Antigen: LGMN
Clonality: Monoclonal
Clone: 312114
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR