Anti-CA14 Mouse Monoclonal Antibody [clone: 312908]

Supplier: R & D Systems
MAB2195
220019-669EA 561.11 USD
220019-669
Anti-CA14 Mouse Monoclonal Antibody [clone: 312908]
Antibodies
Type: Primary
Antigen: CA14
Clonality: Monoclonal
Clone: 312908
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR