Anti-CA10 Mouse Monoclonal Antibody [clone: 862319]

Supplier: R & D Systems
MAB2189
102885-444EA 594.37 USD
102885-444
Anti-CA10 Mouse Monoclonal Antibody [clone: 862319]
Antibodies
Type: Primary
Antigen: CA10
Clonality: Monoclonal
Clone: 862319
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR