Anti-CA9 Mouse Monoclonal Antibody [clone: 303104]

Supplier: R & D Systems
MAB21881
102885-442EA 577.02 USD
102885-442
Anti-CA9 Mouse Monoclonal Antibody [clone: 303104]
Antibodies
Type: Primary
Antigen: CA9
Clonality: Monoclonal
Clone: 303104
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR