Anti-CA8 Mouse Monoclonal Antibody [clone: 308320]

Supplier: R & D Systems
MAB2187
220019-665EA 566.89 USD
220019-665
Anti-CA8 Mouse Monoclonal Antibody [clone: 308320]
Antibodies
Type: Primary
Antigen: CA8
Clonality: Monoclonal
Clone: 308320
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR