Anti-CA4 Mouse Monoclonal Antibody [clone: 310415]

Supplier: R & D Systems
MAB21861
220019-664EA 594.37 USD
220019-664
Anti-CA4 Mouse Monoclonal Antibody [clone: 310415]
Antibodies
Type: Primary
Antigen: CA4
Clonality: Monoclonal
Clone: 310415
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR