Anti-CA4 Mouse Monoclonal Antibody [clone: 310413]

Supplier: R & D Systems
MAB2186
220019-663EA 594.37 USD
220019-663
Anti-CA4 Mouse Monoclonal Antibody [clone: 310413]
Antibodies
Type: Primary
Antigen: CA4
Clonality: Monoclonal
Clone: 310413
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR